Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256604.1 | NP_001243533.1 | 1 | 114 | ||
NM_001256605.1 | NP_001243534.1 | 1 | 114 | |||
NM_001256608.1 | NP_001243537.1 | 1 | 114 | |||
NM_001256609.1 | NP_001243538.1 | 1 | 114 | |||
NM_144580.2 | NP_653181.1 | 1 | 114 | |||
XM_017000149.1 | XP_016855638.1 | 1 | 110 | |||
GenBank mRNA | AK075349.1 | - | 1 | 83 | ||
AK296157.1 | - | 1 | 82 | |||
AL528533.3 | - | 1 | 86 | |||
AY358450.1 | - | 1 | 87 | |||
BC011575.1 | - | 1 | 97 | |||
BC018757.1 | - | 1 | 83 | 4850051 | ||
BC036340.1 | - | 1 | 30 | |||
CX756681.1 | - | 1 | 29 | |||
DA537642.1 | - | 1 | 114 |