Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052950.3 | NP_443182.1 | 11 | 1476 | ||
XM_011534914.1 | XP_011533216.1 | 11 | 1367 | |||
XM_011534915.2 | XP_011533217.1 | 10 | 1556 | |||
XM_017020386.1 | XP_016875875.1 | 11 | 7280 | |||
GenBank mRNA | AF411978.1 | - | 11 | 1136 | ||
AK058149.1 | - | 9 | 922 | |||
AK093224.1 | - | 2 | 1012 | |||
BC014004.2 | - | 11 | 1316 | 3842589 |