Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052971.2 | NP_443203.1 | 2 | 197 | ||
GenBank mRNA | AJ306405.1 | - | 2 | 197 | ||
AJ409065.1 | - | 2 | 1541 | |||
AY052782.1 | - | 2 | 197 | |||
BC070199.1 | - | 2 | 195 | 4711742 | ||
BX443829.2 | - | 2 | 185 |