Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030649.2 | NP_085152.2 | 3, 4 | 302 | ||
XM_005244715.2 | XP_005244772.1 | 3, 4 | 338 | |||
XM_017000233.1 | XP_016855722.1 | 3, 4 | 253 | |||
GenBank mRNA | AB051503.1 | - | 2, 3 | 1967 | ||
AF411981.1 | - | 1, 2 | 102 | |||
AK093954.1 | - | 3, 4 | 349 | |||
AK098490.1 | - | 2, 3 | 2506 | |||
BC051194.1 | - | 3, 4 | 302 | 4819254 |