Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152309.2 | NP_689522.2 | 6 | 1010 | ||
XM_005269498.1 | XP_005269555.1 | 5 | 533 | |||
XM_005269499.1 | XP_005269556.1 | 5 | 557 | |||
XM_011539248.1 | XP_011537550.1 | 6 | 1002 | |||
XM_011539249.1 | XP_011537551.1 | 6 | 1002 | |||
GenBank mRNA | AK092883.1 | - | 5 | 533 | ||
AK125635.1 | - | 6 | 1002 | |||
BX648550.1 | - | 6 | 906 |