Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003700.1 | NP_003691.1 | 1 | 122 | ||
GenBank mRNA | BC140729.1 | - | 1 | 221 | 9021597 |