Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193380.1 | NP_001180309.1 | 11, 12 | 1187 | ||
NM_153480.1 | NP_705613.1 | 11, 12 | 1187 | |||
NM_153481.1 | NP_705614.1 | 9, 10 | 1051 | |||
NM_153483.2 | NP_705616.2 | 12, 13 | 1206 | |||
NR_104198.1 | 10, 11 | 1171 | ||||
XM_006712975.1 | XP_006713038.1 | 10, 11 | 1013 | |||
XM_006712976.1 | XP_006713039.1 | 10, 11 | 1151 | |||
XM_011533361.2 | XP_011531663.1 | 11, 12 | 1103 | |||
XM_011533363.2 | XP_011531665.1 | 11, 12 | 1122 | |||
GenBank mRNA | AF458069.1 | - | 11, 12 | 1187 | ||
AK074238.1 | - | 11, 12 | 1167 | |||
AK096793.1 | - | 10, 11 | 1149 | |||
AK096801.1 | - | 9, 10 | 1031 | |||
AK297697.1 | - | 11, 12 | 1153 | |||
AK298025.1 | - | 7, 8 | 866 | |||
AK314476.1 | - | 11, 12 | 1172 | |||
AY358451.1 | - | 11, 12 | 1187 | |||
BC035478.1 | - | 12, 13 | 1199 | |||
BC137077.1 | - | 12, 13 | 1149 | 9021074 | ||
BC137078.1 | - | 12, 13 | 1149 | 9021075 |