Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A2ML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282424.2 | NP_001269353.1 | 23, 24 | 3022 | ||
NM_144670.5 | NP_653271.2 | 34, 35 | 4507 | |||
XM_011520566.2 | XP_011518868.1 | 34, 35 | 4464 | |||
XM_011520567.2 | XP_011518869.1 | 34, 35 | 4464 | |||
XM_017018868.1 | XP_016874357.1 | 34, 35 | 4461 | |||
XM_017018869.1 | XP_016874358.1 | 34, 35 | 4461 | |||
XM_017018870.1 | XP_016874359.1 | 34, 35 | 4425 | |||
GenBank mRNA | AK057908.1 | - | 5, 6 | 509 | ||
AK096448.1 | - | 12, 13 | 1496 | |||
AK122624.1 | - | 28, 29 | 3593 | |||
AK123591.1 | - | 17, 18 | 2160 | |||
AK123592.1 | - | 23, 24 | 2977 | |||
AK123601.1 | - | 31, 32 | 3884 | |||
AK290355.1 | - | 34, 35 | 4360 | |||
AK302555.1 | - | 23, 24 | 3022 | |||
AL832139.1 | - | 34, 35 | 4358 | |||
AL832750.1 | - | 12, 13 | 1232 | |||
AL833479.1 | - | 32, 33 | 4023 | |||
BC093840.1 | - | 5, 6 | 460 | |||
BC112131.1 | - | 5, 6 | 460 | |||
BX647674.1 | - | 16, 17 | 1962 | |||
BX648231.1 | - | 35, 36 | 4359 |