Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287010.1 | NP_001273939.1 | 4 | 585 | ||
NM_001287011.1 | NP_001273940.1 | 5 | 793 | |||
NM_001287012.1 | NP_001273941.1 | 3 | 481 | |||
NM_001287013.1 | NP_001273942.1 | 5 | 500 | |||
NM_001287014.1 | NP_001273943.1 | 2 | 209 | |||
NM_022343.3 | NP_071738.1 | 5 | 663 | |||
NR_104637.1 | 7 | 1044 | ||||
NR_104638.1 | 5 | 455 | ||||
NR_104639.1 | 6 | 798 | ||||
NR_104640.1 | 5 | 685 | ||||
NR_104641.1 | 2 | 372 | ||||
GenBank mRNA | AJ011129.1 | - | 4 | 420 | ||
AK075091.1 | - | 5 | 464 | |||
AK091288.1 | - | 6 | 1277 | |||
AK307243.1 | - | 5 | 594 | |||
AY039756.1 | - | 5 | 454 | |||
CR749781.1 | - | 7 | 845 | |||
DA268281.1 | - | 5 | 500 | |||
DA271525.1 | - | 5 | 440 | |||
DA432700.1 | - | 3 | 282 | |||
DA824243.1 | - | 5 | 454 |