Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHAL6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144071.1 | NP_001137543.1 | 3, 4 | 494 | ||
NM_144972.4 | NP_659409.2 | 2, 3 | 1087 | |||
XM_005252805.4 | XP_005252862.2 | 3, 4 | 376 | |||
XM_011519922.2 | XP_011518224.1 | 3, 4 | 378 | |||
XM_011519923.2 | XP_011518225.1 | 3, 4 | 465 | |||
XM_011519924.2 | XP_011518226.1 | 3, 4 | 322 | |||
XM_011519925.1 | XP_011518227.1 | 3, 4 | 304 | |||
GenBank mRNA | AK131523.1 | - | 2, 3 | 1030 | ||
AY581313.1 | - | 3, 4 | 494 | |||
BC014340.1 | - | 3, 4 | 498 | 3935569 |