Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SASS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304829.1 | NP_001291758.1 | 15 | 2048 | ||
NM_194292.2 | NP_919268.1 | 16 | 2128 | |||
XM_017000486.1 | XP_016855975.1 | 16 | 1894 | |||
GenBank mRNA | AK025750.1 | - | 4 | 426 | ||
AK302513.1 | - | 17 | 1878 | |||
AK307824.1 | - | 15 | 1914 | |||
AL834265.1 | - | 12 | 1391 | |||
AY359522.1 | - | 16 | 1988 | |||
BC101026.3 | - | 15 | 1841 | 40007432 | ||
BX641109.1 | - | 15 | 2039 | |||
BX648406.1 | - | 16 | 1885 |