Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142497.2 | NP_001135969.1 | 2 | 1065 | ||
NM_152649.3 | NP_689862.1 | 2 | 1065 | |||
XM_005255834.3 | XP_005255891.1 | 3 | 964 | |||
XM_011522936.2 | XP_011521238.1 | 2 | 1104 | |||
GenBank mRNA | AK091708.1 | - | 2 | 808 | ||
AL833362.1 | - | 2 | 482 | |||
BC028141.1 | - | 2 | 707 | 5214733 |