Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256088.1 | NP_001243017.1 | 2 | 240 | ||
NM_145013.2 | NP_659450.1 | 2 | 229 | |||
NR_045767.1 | 2 | 163 | ||||
XM_011542654.2 | XP_011540956.1 | 2 | 159 | |||
XM_011542655.2 | XP_011540957.1 | 2 | 121 | |||
GenBank mRNA | AK125634.1 | - | 2 | 137 | ||
AK314137.1 | - | 2 | 206 | |||
BC025756.1 | - | 2 | 229 | 5201867 | ||
BX956612.1 | - | 2 | 163 | |||
DB219954.1 | - | 2 | 242 |