Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSBN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198467.2 | NP_940869.2 | 8 | 2212 | ||
GenBank mRNA | AK124517.1 | - | 4 | 579 | ||
AK127713.1 | - | 6 | 1407 | |||
BC046193.1 | - | 1 | 155 | |||
BC059402.1 | - | 8 | 2183 | 30347338 |