Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC57 SIRNA Assays ›
Gene Symbol:
CCDC57
Gene Name:
coiled-coil domain containing 57
Gene Aliases:
-
Chromosome Location:
Chr.17: 82101458 - 82212892 on Build GRCh38
UniGene ID:
Hs.631724
Species:
Human
siRNA ID
HSS138448
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC57
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001316321.1
NP_001303250.1
13
2131
NM_198082.2
NP_932348.2
12
1797
XM_011523545.2
XP_011521847.2
13
2034
XM_011523546.2
XP_011521848.1
12
2366
XM_011523548.2
XP_011521850.1
13
2062
XM_011523549.2
XP_011521851.1
12
1876
XM_011523551.2
XP_011521853.1
13
2075
XM_011523552.2
XP_011521854.1
11
1434
XM_011523555.2
XP_011521857.1
10
2434
XM_011523556.2
XP_011521858.2
13
2028
XM_011523557.2
XP_011521859.1
6
798
XM_017024462.1
XP_016879951.1
13
2036
XM_017024463.1
XP_016879952.1
13
2025
XM_017024464.1
XP_016879953.1
13
2027
XM_017024465.1
XP_016879954.1
13
2026
XM_017024466.1
XP_016879955.1
13
2015
XM_017024467.1
XP_016879956.1
13
2191
XM_017024468.1
XP_016879957.1
13
2022
XM_017024469.1
XP_016879958.1
13
1854
XM_017024470.1
XP_016879959.1
12
1655
XM_017024471.1
XP_016879960.1
12
1831
XM_017024472.1
XP_016879961.1
13
2052
XM_017024473.1
XP_016879962.1
13
2021
XM_017024474.1
XP_016879963.1
13
2025
XM_017024475.1
XP_016879964.1
13
1761
XM_017024476.1
XP_016879965.1
11
1937
XM_017024477.1
XP_016879966.1
12
2105
XM_017024478.1
XP_016879967.1
10
1568
XM_017024479.1
XP_016879968.1
12
2241
XM_017024483.1
XP_016879972.1
10
1670
GenBank mRNA
AK074059.1
-
5
588
BC040264.1
-
13
2115
5165045
BC110997.1
-
12
1797
5725904
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-c74kb:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0