Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213604.2 | NP_998769.2 | 2 | 486 | ||
XM_005259549.3 | XP_005259606.1 | 3 | 195 | |||
XM_011527961.1 | XP_011526263.1 | 3 | 195 | |||
XM_011527963.1 | XP_011526265.1 | 3 | 191 | |||
XM_017026744.1 | XP_016882233.1 | 2 | 317 | |||
XM_017026745.1 | XP_016882234.1 | 2 | 349 | |||
GenBank mRNA | AK131571.1 | - | 2 | 317 | ||
AK302020.1 | - | 3 | 221 | |||
BC040620.1 | - | 2 | 486 | |||
BC119796.1 | - | 2 | 117 | |||
BC139833.1 | - | 2 | 77 | 40033483 |