Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201628.2 | NP_963922.2 | 14 | 2420 | ||
XM_005245795.4 | XP_005245852.1 | 16 | 2675 | |||
XM_011541074.2 | XP_011539376.1 | 16 | 2682 | |||
XM_011541075.2 | XP_011539377.1 | 14 | 2338 | |||
XM_011541076.2 | XP_011539378.1 | 14 | 2483 | |||
XM_011541077.2 | XP_011539379.1 | 14 | 3462 | |||
XM_011541081.2 | XP_011539383.1 | 8 | 1162 | |||
GenBank mRNA | AK293806.1 | - | 9 | 1226 | ||
BC101637.1 | - | 4 | 501 | |||
BC113621.1 | - | 4 | 501 | |||
BC143641.1 | - | 4 | 447 | |||
DA317442.1 | - | 4 | 438 | |||
DN831635.1 | - | 5 | 702 |
Gene Symbol: TMEM51-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027136.1 | 5 | 6273 |