Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164664.1 | NP_001158136.1 | 20 | 2871 | ||
NM_001290226.1 | NP_001277155.1 | 18 | 2323 | |||
NM_001290227.1 | NP_001277156.1 | 18 | 2095 | |||
NM_001297651.1 | NP_001284580.1 | 17 | 2355 | |||
NM_015183.2 | NP_055998.1 | 19 | 2064 | |||
XM_006714606.3 | XP_006714669.1 | 18 | 2317 | |||
XM_006714610.2 | XP_006714673.1 | 17 | 2304 | |||
XM_011543382.2 | XP_011541684.1 | 22 | 2963 | |||
XM_011543384.2 | XP_011541686.1 | 17 | 4022 | |||
XM_011543385.2 | XP_011541687.1 | 17 | 5142 | |||
XM_011543386.2 | XP_011541688.1 | 17 | 1973 | |||
XM_017009447.1 | XP_016864936.1 | 23 | 3044 | |||
XM_017009448.1 | XP_016864937.1 | 21 | 2927 | |||
XM_017009449.1 | XP_016864938.1 | 18 | 2362 | |||
XM_017009450.1 | XP_016864939.1 | 18 | 2056 | |||
XM_017009451.1 | XP_016864940.1 | 18 | 2056 | |||
XM_017009452.1 | XP_016864941.1 | 18 | 5371 | |||
XM_017009453.1 | XP_016864942.1 | 17 | 12186 | |||
GenBank mRNA | AB002301.1 | - | 9 | 1107 | ||
AK131421.1 | - | 19 | 2403 | |||
AK296707.1 | - | 18 | 2060 | |||
AK299538.1 | - | 18 | 2205 | |||
AK299706.1 | - | 17 | 2354 | |||
AY830839.1 | - | 19 | 2065 |