Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDK10 SIRNA Assays ›
Gene Symbol:
CDK10
Gene Name:
cyclin dependent kinase 10
Gene Aliases:
PISSLRE
Chromosome Location:
Chr.16: 89686612 - 89696364 on Build GRCh38
UniGene ID:
Hs.699177
Species:
Human
siRNA ID
HSS140783
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098533.2
NP_001092003.2
6
468
NM_001160367.1
NP_001153839.1
6
468
NM_052987.3
NP_443713.2
6
468
NM_052988.4
NP_443714.3
6
506
NR_027702.1
5
433
NR_027703.1
5
433
XM_006721308.2
XP_006721371.1
6
562
XM_006721310.2
XP_006721373.1
6
562
XM_011523405.2
XP_011521707.1
6
562
XM_011523406.2
XP_011521708.1
6
562
XM_011523407.2
XP_011521709.1
6
562
XM_011523408.2
XP_011521710.1
6
562
XM_011523410.2
XP_011521712.1
6
480
XM_011523411.2
XP_011521713.1
7
718
XM_011523412.1
XP_011521714.1
7
688
XM_011523413.2
XP_011521715.1
6
1120
XM_011523414.2
XP_011521716.1
6
906
XM_011523415.2
XP_011521717.1
5
437
XM_011523416.2
XP_011521718.1
6
645
XM_011523417.2
XP_011521719.1
6
562
XM_017023806.1
XP_016879295.1
5
451
XM_017023807.1
XP_016879296.1
6
562
XM_017023808.1
XP_016879297.1
7
717
XM_017023809.1
XP_016879298.1
5
437
XM_017023810.1
XP_016879299.1
6
905
GenBank mRNA
AB209829.1
-
5
413
AK131531.1
-
5
2065
AK290485.1
-
6
426
AK292351.1
-
6
484
AK295397.1
-
6
472
AK296631.1
-
5
410
AK298386.1
-
6
506
BC017342.2
-
6
448
4865641
BC025301.1
-
5
401
4637860
BC093028.1
-
6
485
CX867290.1
-
6
471
DB118903.1
-
5
433
DC362478.1
-
6
506
L33264.1
-
6
467
X78342.1
-
6
583
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
traversing start control point of mitotic cell cycle
negative regulation of cell proliferation
positive regulation of MAPK cascade
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-jhld4:80/10.239.165.190:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0