Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDK10 SIRNA Assays ›
Gene Symbol:
CDK10
Gene Name:
cyclin dependent kinase 10
Gene Aliases:
PISSLRE
Chromosome Location:
Chr.16: 89686612 - 89696364 on Build GRCh38
UniGene ID:
Hs.699177
Species:
Human
siRNA ID
HSS141079
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098533.2
NP_001092003.2
10
749
NM_001160367.1
NP_001153839.1
10
749
NM_052987.3
NP_443713.2
10
749
NM_052988.4
NP_443714.3
10
787
NR_027702.1
9
714
NR_027703.1
9
714
XM_006721308.2
XP_006721371.1
10
843
XM_006721310.2
XP_006721373.1
10
843
XM_011523405.2
XP_011521707.1
10
939
XM_011523406.2
XP_011521708.1
10
939
XM_011523407.2
XP_011521709.1
10
939
XM_011523408.2
XP_011521710.1
10
939
XM_011523410.2
XP_011521712.1
10
857
XM_011523411.2
XP_011521713.1
11
1095
XM_011523412.1
XP_011521714.1
11
1065
XM_011523413.2
XP_011521715.1
10
1497
XM_011523414.2
XP_011521716.1
10
1283
XM_011523415.2
XP_011521717.1
9
814
XM_011523416.2
XP_011521718.1
10
1022
XM_017023806.1
XP_016879295.1
9
828
XM_017023807.1
XP_016879296.1
10
843
XM_017023808.1
XP_016879297.1
11
998
XM_017023809.1
XP_016879298.1
9
718
XM_017023810.1
XP_016879299.1
10
1186
GenBank mRNA
AB209829.1
-
7
1056
AF153430.1
-
8
754
AK131531.1
-
9
2346
AK296631.1
-
9
691
AK298386.1
-
10
787
BC017342.2
-
10
729
4865641
BC025301.1
-
9
682
4637860
BC093028.1
-
9
837
BM720563.1
-
2
130
CX867290.1
-
10
752
L33264.1
-
10
748
X78342.1
-
10
864
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
traversing start control point of mitotic cell cycle
negative regulation of cell proliferation
positive regulation of MAPK cascade
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-sjrww:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0