Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178029.1 | NP_001171500.1 | 4 | 407 | ||
NM_001919.3 | NP_001910.2 | 4 | 407 | |||
GenBank mRNA | AK291127.1 | - | 4 | 407 | ||
BC000762.2 | - | 4 | 372 | 2821822 | ||
BC002746.2 | - | 4 | 378 | 3630566 | ||
BC009631.1 | - | 5 | 349 | 3877066 | ||
BC019316.1 | - | 4 | 372 | 2821822 | ||
BC020228.1 | - | 4 | 378 | 4541779 | ||
BQ773923.1 | - | 4 | 660 | |||
CD675473.1 | - | 4 | 408 | |||
L24774.1 | - | 3 | 264 | |||
Z25820.1 | - | 4 | 380 |