Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL1 SIRNA Assays ›
Gene Symbol:
MBNL1
Gene Name:
muscleblind like splicing regulator 1
Gene Aliases:
EXP, MBNL
Chromosome Location:
Chr.3: 152243656 - 152465780 on Build GRCh38
UniGene ID:
Hs.201858
Species:
Human
siRNA ID
HSS142884
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001314057.1
NP_001300986.1
6
1311
NM_021038.4
NP_066368.2
5
2096
NM_207292.2
NP_997175.1
5
2096
NM_207293.1
NP_997176.1
4
1471
NM_207294.1
NP_997177.1
3
1267
NM_207295.1
NP_997178.1
3
1267
NM_207296.1
NP_997179.1
4
1471
NM_207297.1
NP_997180.1
4
1471
XM_005247457.4
XP_005247514.1
6
1664
XM_005247458.4
XP_005247515.1
6
2918
XM_005247460.4
XP_005247517.1
6
1627
XM_005247461.4
XP_005247518.1
6
1664
XM_005247462.4
XP_005247519.1
6
1664
XM_005247463.4
XP_005247520.1
6
1664
XM_005247464.4
XP_005247521.1
5
1722
XM_005247465.4
XP_005247522.1
6
1664
XM_005247466.4
XP_005247523.1
5
1699
XM_005247467.4
XP_005247524.1
6
1664
XM_005247468.4
XP_005247525.1
6
1664
XM_005247471.4
XP_005247528.1
5
1694
XM_005247474.3
XP_005247531.1
4
582
XM_005247475.3
XP_005247532.1
5
905
XM_005247476.4
XP_005247533.1
4
672
XM_005247477.4
XP_005247534.1
4
671
XM_006713639.1
XP_006713702.1
6
1646
XM_011512843.2
XP_011511145.1
5
1460
XM_011512845.2
XP_011511147.1
5
1460
XM_011512846.2
XP_011511148.1
5
1460
XM_011512847.2
XP_011511149.1
4
1423
XM_011512848.2
XP_011511150.1
6
1268
XM_011512849.2
XP_011511151.1
4
1367
XM_017006423.1
XP_016861912.1
5
1915
XM_017006424.1
XP_016861913.1
5
1742
XM_017006425.1
XP_016861914.1
5
1744
XM_017006426.1
XP_016861915.1
5
1742
XM_017006427.1
XP_016861916.1
5
1889
XM_017006428.1
XP_016861917.1
5
1553
XM_017006429.1
XP_016861918.1
5
1636
XM_017006430.1
XP_016861919.1
5
1700
XM_017006431.1
XP_016861920.1
5
1777
XM_017006432.1
XP_016861921.1
5
1699
XM_017006433.1
XP_016861922.1
5
1699
XM_017006434.1
XP_016861923.1
5
1705
XM_017006435.1
XP_016861924.1
5
712
XM_017006436.1
XP_016861925.1
5
2825
XM_017006437.1
XP_016861926.1
5
1880
XM_017006438.1
XP_016861927.1
4
1367
XM_017006439.1
XP_016861928.1
5
905
XM_017006440.1
XP_016861929.1
4
639
XM_017006441.1
XP_016861930.1
4
672
XM_017006442.1
XP_016861931.1
4
775
XM_017006443.1
XP_016861932.1
4
783
GenBank mRNA
AB007888.1
-
5
2096
AF255334.1
-
3
989
AF395876.1
-
3
478
AF401998.1
-
4
682
AF497718.1
-
4
682
AF497719.1
-
4
682
AJ308400.1
-
4
711
AK297133.1
-
2
187
AK304089.1
-
3
391
AK307420.1
-
3
391
BC043493.1
-
5
1601
6044671
BC050535.1
-
6
832
6042311
Y13829.1
-
4
1193
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
mRNA processing
nervous system development
RNA splicing
embryonic limb morphogenesis
regulation of RNA splicing
myoblast differentiation
regulatory region RNA binding
RNA binding
double-stranded RNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0