Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002810.2 | NP_002801.1 | 6, 7 | 715 | ||
XM_005245354.2 | XP_005245411.1 | 6, 7 | 736 | |||
XM_006711464.3 | XP_006711527.1 | 5, 6 | 561 | |||
XM_017001856.1 | XP_016857345.1 | 5, 6 | 556 | |||
GenBank mRNA | AB033605.1 | - | 6, 7 | 667 | ||
BC002365.2 | - | 6, 7 | 665 | 2820813 | ||
BC072008.1 | - | 6, 7 | 679 | 6285035 | ||
U24704.1 | - | 6, 7 | 715 | |||
U72664.1 | - | 4, 5 | 447 |