Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000979.3 | NP_000970.1 | 2, 3 | 362 | ||
GenBank mRNA | AK026558.1 | - | 2, 3 | 120 | ||
AK293386.1 | - | 2, 3 | 121 | |||
BC000374.2 | - | 1, 2 | 94 | 2820119 | ||
BC009708.2 | - | 2, 3 | 108 | 3878799 | ||
BC021743.2 | - | 2, 3 | 120 | 5261251 | ||
BM764512.1 | - | 2, 3 | 480 | |||
DQ403032.1 | - | 1, 2 | 18 | |||
L11566.1 | - | 2, 3 | 102 |