Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMTN SIRNA Assays ›
Gene Symbol:
SMTN
Gene Name:
smoothelin
Gene Aliases:
-
Chromosome Location:
Chr.22: 31081296 - 31104624 on Build GRCh38
UniGene ID:
Hs.149098
Species:
Human
siRNA ID
HSS143958
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001207017.1
NP_001193946.1
7
899
NM_001207018.1
NP_001193947.1
6
921
NM_006932.4
NP_008863.3
7
917
NM_134269.2
NP_599031.1
7
917
NM_134270.2
NP_599032.2
7
917
XM_006724298.1
XP_006724361.1
7
895
XM_006724302.1
XP_006724365.1
7
954
XM_011530333.2
XP_011528635.1
8
1000
XM_011530336.2
XP_011528638.1
8
1001
XM_011530337.2
XP_011528639.1
8
1001
XM_011530338.2
XP_011528640.1
8
1000
XM_011530339.2
XP_011528641.1
8
1001
XM_011530340.2
XP_011528642.1
8
1001
XM_017028913.1
XP_016884402.1
8
1001
XM_017028914.1
XP_016884403.1
8
1000
XM_017028915.1
XP_016884404.1
8
1000
XM_017028916.1
XP_016884405.1
8
1001
XM_017028917.1
XP_016884406.1
8
1001
XM_017028918.1
XP_016884407.1
8
1001
XM_017028919.1
XP_016884408.1
8
1000
GenBank mRNA
AB209643.1
-
4
490
AF064238.3
-
7
894
AJ010306.2
-
7
894
AK096953.1
-
7
952
AK128627.1
-
6
2462
AK128719.1
-
7
763
AK295418.1
-
6
921
AK304084.1
-
7
899
AY061971.1
-
7
895
AY061972.1
-
7
895
BC034237.2
-
7
885
3912887
Y13492.2
-
7
895
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
smooth muscle contraction
muscle organ development
actin binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0