Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286416.1 | NP_001273345.1 | 1, 2 | 183 | ||
NM_006936.2 | NP_008867.2 | 1, 2 | 183 | |||
GenBank mRNA | AK222591.1 | - | 1, 2 | 114 | ||
AK300822.1 | - | 1, 2 | 135 | |||
AK312350.1 | - | 1, 2 | 114 | |||
BC000036.2 | - | 1, 2 | 108 | 3505840 | ||
BC008420.1 | - | 1, 2 | 183 | 4091864 | ||
BT007008.1 | - | 1, 2 | 22 | |||
CR542173.1 | - | 1, 2 | 22 | |||
CR542188.1 | - | 1, 2 | 22 | |||
X99584.1 | - | 1, 2 | 116 |