Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005985.3 | NP_005976.2 | 2 | 255 | ||
GenBank mRNA | AF125377.1 | - | 2 | 232 | ||
AF131208.1 | - | 2 | 171 | |||
AK313228.1 | - | 2 | 251 | |||
BC012910.1 | - | 2 | 236 | 4537122 | ||
BI822180.1 | - | 2 | 242 | |||
DA972913.1 | - | 2 | 255 |