Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRR2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014291.3 | NP_001014313.1 | 2 | 233 | ||
XM_017002177.1 | XP_016857666.1 | 4 | 610 | |||
XM_017002178.1 | XP_016857667.1 | 2 | 156 | |||
GenBank mRNA | AA456642.1 | - | 1 | 434 | ||
AA490630.1 | - | 1 | 143 | |||
BC130455.1 | - | 2 | 233 | |||
BC133041.1 | - | 2 | 233 |