Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PQBP1 SIRNA Assays ›
Gene Symbol:
PQBP1
Gene Name:
polyglutamine binding protein 1
Gene Aliases:
MRX2, MRX55, MRXS3, MRXS8, NPW38, RENS1, SHS
Chromosome Location:
Chr.X: 48897862 - 48903145 on Build GRCh38
UniGene ID:
Hs.534384
Species:
Human
siRNA ID
HSS145445
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PQBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001032381.1
NP_001027553.1
3
267
NM_001032382.1
NP_001027554.1
3
335
NM_001032383.1
NP_001027555.1
3
346
NM_001032384.1
NP_001027556.1
3
255
NM_001167989.1
NP_001161461.1
3
267
NM_001167992.1
NP_001161464.1
2
164
NM_005710.2
NP_005701.1
2
400
NM_144495.2
NP_652766.1
3
335
XM_005272571.3
XP_005272628.1
3
330
XM_005272572.4
XP_005272629.1
3
258
XM_011543884.2
XP_011542186.1
3
405
XM_017029207.1
XP_016884696.1
3
396
GenBank mRNA
AB016533.1
-
3
316
AB041833.1
-
2
146
AB041834.1
-
2
146
AB041835.1
-
2
146
AB041836.1
-
2
146
AJ005893.1
-
2
403
AJ242829.1
-
3
206
AJ973593.1
-
2
161
AJ973594.1
-
2
161
AJ973595.1
-
2
161
AJ973596.1
-
161
AJ973597.1
-
2
161
AJ973598.1
-
2
161
AJ973599.1
-
2
161
AJ973600.1
-
2
161
AJ973601.1
-
2
161
AJ973602.1
-
2
161
AJ973603.1
-
2
161
AJ973604.1
-
2
165
AJ973605.1
-
2
161
AJ973606.1
-
2
161
AJ973607.1
-
2
161
BC012358.1
-
3
227
4662649
BE385548.1
-
1
81
BE396796.1
-
3
247
BI818141.1
-
3
229
BX362311.2
-
3
304
BX369697.2
-
3
487
CN292761.1
-
3
335
CN292769.1
-
3
253
KU178500.1
-
2
146
KU178501.1
-
2
146
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
alternative mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
neuron projection development
regulation of RNA splicing
regulation of dendrite morphogenesis
cilium morphogenesis
DNA binding
transcription coactivator activity
protein binding
protein C-terminus binding
ribonucleoprotein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-c74kb:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0