Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001120.4 | NP_001111.3 | 5 | 929 | ||
NM_001146069.1 | NP_001139541.1 | 6 | 727 | |||
XM_005247927.3 | XP_005247984.1 | 6 | 722 | |||
GenBank mRNA | BC001502.2 | - | 6 | 653 | 2988084 | |
BC014979.1 | - | 6 | 656 | 4878602 | ||
L11669.1 | - | 5 | 735 |