Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098816.2 | NP_001092286.2 | 28 | 4698 | ||
XM_011544933.2 | XP_011543235.1 | 11 | 1793 | |||
XM_017017525.1 | XP_016873014.1 | 27 | 4969 | |||
XM_017017526.1 | XP_016873015.1 | 25 | 5874 | |||
GenBank mRNA | AB037723.3 | - | 13 | 1976 | ||
BX640737.1 | - | 2 | 125 |