Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017623.4 | NP_060093.3 | 3, 4 | 1542 | ||
NM_199078.2 | NP_951060.1 | 2, 3 | 1398 | |||
XM_011510957.2 | XP_011509259.1 | 3, 4 | 1902 | |||
XM_017003800.1 | XP_016859289.1 | 3, 4 | 1902 | |||
GenBank mRNA | AF216965.1 | - | 3, 4 | 641 | ||
AK000025.1 | - | 2, 3 | 460 | |||
AK126847.1 | - | 3, 4 | 1542 | |||
BC007199.1 | - | 2, 3 | 554 | 2964706 | ||
BC018604.1 | - | 2, 3 | 287 | |||
BC037272.1 | - | 3, 4 | 1551 | 4823510 | ||
CR749702.1 | - | 2, 3 | 176 |