Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5AR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271749.1 | NP_001258678.1 | 2 | 557 | ||
NM_001271750.1 | NP_001258679.1 | 2 | 480 | |||
NM_018485.2 | NP_060955.1 | 2 | 519 | |||
XM_011526736.2 | XP_011525038.1 | 3 | 1194 | |||
GenBank mRNA | AB038237.1 | - | 2 | 410 | ||
AK313994.1 | - | 2 | 557 | |||
BC067477.1 | - | 1 | 358 | 6971811 | ||
BC067478.1 | - | 1 | 358 | 6971812 | ||
BC067480.1 | - | 1 | 358 | 6971814 | ||
BP370117.1 | - | 2 | 480 | |||
DA673976.1 | - | 2 | 519 |
Set Membership: |
G-protein coupled receptor |