Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IER5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016545.4 | NP_057629.2 | 1 | 1143 | ||
GenBank mRNA | AF178984.1 | - | 1 | 884 | ||
AF258581.1 | - | 1 | 931 | |||
AJ251089.1 | - | 1 | 973 | |||
AK314830.1 | - | 1 | 945 | |||
BC000128.2 | - | 1 | 933 | 3048356 | ||
BC108253.1 | - | 1 | 929 | 6025403 |