Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037585.1 | 3 | 345 | |||
NR_037586.1 | 3 | 345 | ||||
GenBank mRNA | AI420675.1 | - | 2 | 357 | ||
BC130500.1 | - | 3 | 316 | |||
BC130504.1 | - | 3 | 316 | |||
BC144390.1 | - | 3 | 316 | |||
BC144391.1 | - | 3 | 316 | 9052912 | ||
BX089409.1 | - | 3 | 341 | |||
N47348.1 | - | 3 | 342 | |||
N56968.1 | - | 2 | 365 |