Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145265.1 | NP_001138737.1 | 3 | 1367 | ||
NM_001145266.1 | NP_001138738.1 | 3 | 1270 | |||
NM_018389.4 | NP_060859.4 | 2 | 1641 | |||
XM_011520202.2 | XP_011518504.1 | 2 | 528 | |||
GenBank mRNA | AF323970.1 | - | 2 | 929 | ||
AF326199.1 | - | 2 | 1538 | |||
AK000527.1 | - | 1 | 186 | |||
AK002182.1 | - | 3 | 1043 | |||
AK027394.1 | - | 2 | 1618 | |||
AK074929.1 | - | 2 | 1642 | |||
AK091245.1 | - | 2 | 2325 | |||
AK225428.1 | - | 3 | 1065 | |||
AK315473.1 | - | 2 | 1343 | |||
BC001427.2 | - | 2 | 946 | 3139599 |