Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020179.2 | NP_064564.1 | 3 | 388 | ||
XM_011542907.1 | XP_011541209.1 | 3 | 413 | |||
XM_011542908.2 | XP_011541210.1 | 4 | 516 | |||
XM_011542909.1 | XP_011541211.1 | 3 | 398 | |||
XM_011542910.2 | XP_011541212.1 | 3 | 1151 | |||
XM_011542911.2 | XP_011541213.1 | 3 | 1077 | |||
XM_017018019.1 | XP_016873508.1 | 3 | 1133 | |||
XM_017018020.1 | XP_016873509.1 | 4 | 574 | |||
GenBank mRNA | AF197137.1 | - | 3 | 360 | ||
BC031564.1 | - | 3 | 388 | 5189479 |