Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080428.2 | NP_001073897.2 | 16 | 3179 | ||
NM_001122679.1 | NP_001116151.1 | 19 | 3730 | |||
XM_005265950.2 | XP_005266007.1 | 21 | 4377 | |||
XM_005265952.2 | XP_005266009.1 | 17 | 3104 | |||
XM_006714897.2 | XP_006714960.1 | 21 | 4377 | |||
XM_011534604.2 | XP_011532906.1 | 20 | 4191 | |||
XM_017009660.1 | XP_016865149.1 | 21 | 4404 | |||
XM_017009661.1 | XP_016865150.1 | 21 | 4404 | |||
XM_017009662.1 | XP_016865151.1 | 20 | 4194 | |||
XM_017009663.1 | XP_016865152.1 | 18 | 3599 | |||
XM_017009664.1 | XP_016865153.1 | 18 | 3512 | |||
XM_017009665.1 | XP_016865154.1 | 18 | 3308 | |||
XM_017009666.1 | XP_016865155.1 | 18 | 3308 | |||
XM_017009667.1 | XP_016865156.1 | 18 | 3281 | |||
XM_017009668.1 | XP_016865157.1 | 18 | 4818 | |||
XM_017009669.1 | XP_016865158.1 | 18 | 4650 | |||
GenBank mRNA | AB032953.2 | - | 10 | 1867 | ||
BX648178.1 | - | 15 | 2773 | |||
CR936703.1 | - | 16 | 3112 |