Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRAMD1B SIRNA Assays ›
Gene Symbol:
GRAMD1B
Gene Name:
GRAM domain containing 1B
Gene Aliases:
-
Chromosome Location:
Chr.11: 123358299 - 123627771 on Build GRCh38
UniGene ID:
Hs.144725
Species:
Human
siRNA ID
HSS148495
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRAMD1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286563.1
NP_001273492.1
16
2157
NM_001286564.1
NP_001273493.1
15
1866
NM_020716.2
NP_065767.1
15
2136
XM_005271619.4
XP_005271676.1
15
2136
XM_005271621.4
XP_005271678.1
16
6523
XM_005271623.4
XP_005271680.1
15
1901
XM_005271624.4
XP_005271681.2
15
1816
XM_006718892.3
XP_006718955.1
16
2053
XM_011542926.2
XP_011541228.1
16
2050
XM_011542929.2
XP_011541231.1
15
2026
XM_011542930.2
XP_011541232.1
15
2667
XM_011542931.2
XP_011541233.1
15
2581
XM_011542933.1
XP_011541235.1
13
2278
XM_017018034.1
XP_016873523.1
17
2121
XM_017018035.1
XP_016873524.1
17
2118
XM_017018036.1
XP_016873525.1
16
2588
XM_017018037.1
XP_016873526.1
17
2887
XM_017018038.1
XP_016873527.1
17
2121
XM_017018039.1
XP_016873528.1
16
2179
XM_017018040.1
XP_016873529.1
15
1975
XM_017018041.1
XP_016873530.1
16
2179
XM_017018042.1
XP_016873531.1
16
2179
XM_017018043.1
XP_016873532.1
15
1812
XM_017018044.1
XP_016873533.1
15
2136
XM_017018045.1
XP_016873534.1
16
6520
XM_017018046.1
XP_016873535.1
16
2625
XM_017018047.1
XP_016873536.1
16
2622
XM_017018048.1
XP_016873537.1
15
2578
XM_017018049.1
XP_016873538.1
16
2619
XM_017018050.1
XP_016873539.1
15
1895
XM_017018051.1
XP_016873540.1
13
1499
GenBank mRNA
AB033027.1
-
15
1799
AK127457.1
-
16
2157
AK297606.1
-
15
1866
AY358924.1
-
5
1389
BC105741.1
-
15
1950
40026783
BC105931.1
-
15
1950
40026781
BC105932.1
-
15
1950
40026782
BC107480.1
-
15
1950
40026784
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0