Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGIRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135053.1 | NP_001128525.1 | 3 | 346 | ||
NM_001135054.1 | NP_001128526.1 | 3 | 419 | |||
NM_021805.2 | NP_068577.2 | 3 | 315 | |||
XM_005253044.2 | XP_005253101.1 | 3 | 419 | |||
XM_005253045.1 | XP_005253102.1 | 3 | 315 | |||
XM_005253046.1 | XP_005253103.1 | 3 | 403 | |||
XM_017018099.1 | XP_016873588.1 | 3 | 419 | |||
XM_017018100.1 | XP_016873589.1 | 3 | 416 | |||
GenBank mRNA | AK025099.1 | - | 3 | 315 | ||
AK172830.1 | - | 3 | 315 | |||
AY358342.1 | - | 2 | 192 | |||
BC003591.1 | - | 2 | 245 | 2821373 | ||
BC025953.1 | - | 3 | 274 | 3939162 | ||
BC106007.1 | - | 3 | 396 | 6092503 | ||
CR457338.1 | - | 1 | 89 | |||
DA963481.1 | - | 3 | 403 | |||
DB112269.1 | - | 3 | 419 |