Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM192A SIRNA Assays ›
Gene Symbol:
FAM192A
Gene Name:
family with sequence similarity 192 member A
Gene Aliases:
C16orf94, CDA018, CDA10, NIP30
Chromosome Location:
Chr.16: 57152466 - 57186242 on Build GRCh38
UniGene ID:
Hs.396740
Species:
Human
siRNA ID
HSS149370
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM192A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024946.2
NP_079222.1
7
876
XM_005256156.4
XP_005256213.1
7
844
XM_005256158.3
XP_005256215.1
7
1931
XM_005256160.2
XP_005256217.1
7
848
XM_005256163.2
XP_005256220.1
8
985
XM_005256164.3
XP_005256221.1
8
930
XM_005256165.2
XP_005256222.1
7
905
XM_006721275.3
XP_006721338.2
7
1614
XM_011523343.2
XP_011521645.1
7
722
XM_017023681.1
XP_016879170.1
7
940
XM_017023682.1
XP_016879171.1
8
2049
XM_017023683.1
XP_016879172.1
8
2000
XM_017023684.1
XP_016879173.1
8
1215
XM_017023685.1
XP_016879174.1
8
1964
XM_017023686.1
XP_016879175.1
9
2034
XM_017023687.1
XP_016879176.1
9
2009
XM_017023688.1
XP_016879177.1
9
1952
XM_017023689.1
XP_016879178.1
9
1333
XM_017023690.1
XP_016879179.1
9
1284
XM_017023691.1
XP_016879180.1
9
1337
XM_017023692.1
XP_016879181.1
8
1121
XM_017023693.1
XP_016879182.1
8
1024
XM_017023694.1
XP_016879183.1
9
1127
XM_017023695.1
XP_016879184.1
9
1239
XM_017023696.1
XP_016879185.1
8
1622
XM_017023697.1
XP_016879186.1
8
1007
XM_017023698.1
XP_016879187.1
8
1730
XM_017023699.1
XP_016879188.1
8
1258
XM_017023700.1
XP_016879189.1
8
727
XM_017023701.1
XP_016879190.1
8
1193
XM_017023702.1
XP_016879191.1
8
913
XM_017023703.1
XP_016879192.1
7
653
XM_017023704.1
XP_016879193.1
7
651
GenBank mRNA
AF212249.1
-
7
829
AF271785.1
-
7
829
AF356585.1
-
6
627
AK023291.1
-
7
776
AK025452.1
-
7
851
AL834421.1
-
7
688
BC020536.1
-
7
844
3849620
BC063409.1
-
7
694
BC071952.1
-
6
645
6305656
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0