Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308207.1 | NP_001295136.1 | 4 | 996 | ||
NM_001308208.1 | NP_001295137.1 | 3 | 649 | |||
NM_025251.2 | NP_079527.1 | 2 | 522 | |||
XM_011517308.1 | XP_011515610.1 | 3 | 803 | |||
XM_011517309.1 | XP_011515611.1 | 3 | 707 | |||
XM_017013870.1 | XP_016869359.1 | 4 | 1875 | |||
XM_017013871.1 | XP_016869360.1 | 3 | 934 | |||
GenBank mRNA | AB051475.1 | - | 4 | 1000 | ||
AK296353.1 | - | 3 | 646 |