Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030572.3 | NP_085049.1 | 5 | 493 | ||
NR_135187.1 | 2 | 424 | ||||
NR_135188.1 | 2 | 296 | ||||
GenBank mRNA | AK027273.1 | - | 5 | 331 | ||
AK075342.1 | - | 5 | 447 | |||
AK311147.1 | - | 2 | 194 | |||
AK311217.1 | - | 2 | 285 | |||
BC004336.1 | - | 5 | 428 | 3631700 | ||
DA071522.1 | - | 2 | 297 | |||
DA260291.1 | - | 2 | 424 | |||
DA380664.1 | - | 5 | 492 |