Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146316.1 | NP_001139788.1 | 2 | 654 | ||
NM_001319670.1 | NP_001306599.1 | 1 | 954 | |||
NM_032641.3 | NP_116030.1 | 2 | 706 | |||
GenBank mRNA | AF403027.1 | - | 1 | 498 | ||
AK297963.1 | - | 2 | 649 | |||
AK310353.1 | - | 1 | 903 | |||
BC002983.1 | - | 2 | 637 | 3546861 | ||
BC071933.1 | - | 2 | 654 | 6302511 |