Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPND | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159846.2 | NP_001153318.1 | 8 | 1119 | ||
NM_001300862.1 | NP_001287791.1 | 10 | 1269 | |||
NM_032868.5 | NP_116257.2 | 10 | 1269 | |||
GenBank mRNA | AK027887.1 | - | 8 | 1092 | ||
AK308468.1 | - | 10 | 1230 | |||
BC032652.1 | - | 10 | 1226 | 5503607 | ||
CR977937.1 | - | 7 | 606 |