Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128225.2 | NP_001121697.1 | 10 | 1261 | ||
NM_152264.4 | NP_689477.2 | 10 | 1261 | |||
NR_134854.1 | 10 | 1326 | ||||
XM_006718381.3 | XP_006718444.1 | 10 | 1145 | |||
XM_006718383.3 | XP_006718446.1 | 9 | 1037 | |||
XM_006718384.2 | XP_006718447.1 | 10 | 1141 | |||
XM_006718385.2 | XP_006718448.1 | 9 | 1033 | |||
XM_011520466.1 | XP_011518768.1 | 10 | 1166 | |||
XM_011520467.1 | XP_011518769.1 | 10 | 1495 | |||
XM_011520468.2 | XP_011518770.1 | 10 | 1588 | |||
XM_011520469.1 | XP_011518771.1 | 9 | 1058 | |||
XM_011520470.1 | XP_011518772.1 | 9 | 1032 | |||
XM_017018540.1 | XP_016874029.1 | 10 | 1119 | |||
XM_017018541.1 | XP_016874030.1 | 9 | 1011 | |||
XM_017018542.1 | XP_016874031.1 | 10 | 1115 | |||
GenBank mRNA | AK098651.1 | - | 10 | 1326 | ||
AL133581.1 | - | 3 | 233 | |||
BC008853.2 | - | 10 | 1127 | 4100291 | ||
BC019016.2 | - | 10 | 1125 | 4100057 | ||
BX647491.1 | - | 8 | 2742 |