Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271907.1 | NP_001258836.1 | 3 | 500 | ||
NM_001271908.1 | NP_001258837.1 | 3 | 418 | |||
NM_001271909.1 | NP_001258838.1 | 2 | 556 | |||
NM_001271910.1 | NP_001258839.1 | 3 | 498 | |||
NM_153025.2 | NP_694570.1 | 3 | 447 | |||
NR_073515.1 | 2 | 587 | ||||
GenBank mRNA | AK056168.1 | - | 3 | 389 | ||
AK125673.1 | - | 2 | 328 | |||
AK128803.1 | - | 2 | 562 | |||
AK303024.1 | - | 3 | 466 | |||
BC148299.2 | - | 3 | 389 | 40112476 | ||
DA300380.1 | - | 3 | 410 | |||
DB025192.1 | - | 3 | 447 | |||
DC357681.1 | - | 3 | 456 | |||
DC374690.1 | - | 3 | 468 |