Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080725.2 | NP_542763.1 | 2 | 1915 | ||
GenBank mRNA | AK098418.1 | - | 2 | 810 | ||
AK127435.1 | - | 2 | 1760 | |||
AL833944.1 | - | 2 | 1791 | |||
AY427953.1 | - | 2 | 810 | |||
BC017001.1 | - | 2 | 1710 | |||
BC032604.1 | - | 2 | 1746 | 5535883 | ||
BC047707.1 | - | 2 | 1735 | 5735551 |