Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIB2 SIRNA Assays ›
Gene Symbol:
MIB2
Gene Name:
mindbomb E3 ubiquitin protein ligase 2
Gene Aliases:
ZZANK1, ZZZ5
Chromosome Location:
Chr.1: 1615188 - 1630610 on Build GRCh38
UniGene ID:
Hs.135805
Species:
Human
siRNA ID
HSS152025
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170686.1
NP_001164157.1
10
1510
NM_001170687.1
NP_001164158.1
10
1480
NM_001170688.1
NP_001164159.1
9
1327
NM_001170689.1
NP_001164160.1
9
1302
NM_080875.2
NP_543151.2
10
1522
NR_033183.1
10
1305
XM_006710372.1
XP_006710435.1
9
1386
XM_011540731.2
XP_011539033.1
11
2259
XM_011540736.2
XP_011539038.1
11
2259
XM_011540737.2
XP_011539039.1
10
2196
XM_011540741.2
XP_011539043.1
9
2091
XM_011540742.2
XP_011539044.1
9
2088
XM_017000349.1
XP_016855838.1
11
2259
XM_017000350.1
XP_016855839.1
11
2247
XM_017000351.1
XP_016855840.1
11
2241
XM_017000352.1
XP_016855841.1
11
2217
XM_017000353.1
XP_016855842.1
10
2196
XM_017000354.1
XP_016855843.1
11
2259
XM_017000355.1
XP_016855844.1
9
2091
XM_017000356.1
XP_016855845.1
9
2088
XM_017000357.1
XP_016855846.1
10
2064
XM_017000358.1
XP_016855847.1
11
1557
XM_017000359.1
XP_016855848.1
11
1545
XM_017000360.1
XP_016855849.1
11
1504
XM_017000361.1
XP_016855850.1
10
1494
XM_017000362.1
XP_016855851.1
11
1597
XM_017000363.1
XP_016855852.1
11
1560
XM_017000364.1
XP_016855853.1
9
1299
XM_017000365.1
XP_016855854.1
11
1287
XM_017000366.1
XP_016855855.1
10
1224
GenBank mRNA
AB074480.1
-
10
1433
AB076691.1
-
10
1391
AB076692.1
-
9
1238
AB076693.1
-
10
1421
AB097000.1
-
10
1356
AK091610.1
-
4
326
AK095914.1
-
4
328
AK096295.1
-
5
433
AK097106.1
-
6
752
AK098785.1
-
10
1391
AK122863.1
-
10
1423
AK128167.1
-
9
1262
AK128553.1
-
9
2436
AK293642.1
-
9
1502
AK302757.1
-
10
1474
AK309362.1
-
8
1392
AL834527.1
-
5
554
BC016490.1
-
10
1359
BC037542.1
-
10
1402
5168033
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0