Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRIMPOL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300768.1 | NP_001287697.1 | 3, 4 | 613 | ||
NM_152683.3 | NP_689896.1 | 3, 4 | 613 | |||
XM_011531719.1 | XP_011530021.1 | 3, 4 | 490 | |||
XM_011531720.1 | XP_011530022.1 | 3, 4 | 528 | |||
XM_011531721.2 | XP_011530023.1 | 3, 4 | 490 | |||
XM_011531724.2 | XP_011530026.1 | 3, 4 | 550 | |||
XM_011531725.1 | XP_011530027.1 | 3, 4 | 464 | |||
XM_011531729.1 | XP_011530031.1 | 3, 4 | 449 | |||
XM_011531730.2 | XP_011530032.1 | 3, 4 | 464 | |||
XM_017007864.1 | XP_016863353.1 | 3, 4 | 528 | |||
XM_017007865.1 | XP_016863354.1 | 3, 4 | 528 | |||
XM_017007866.1 | XP_016863355.1 | 3, 4 | 528 | |||
XM_017007867.1 | XP_016863356.1 | 3, 4 | 490 | |||
XM_017007868.1 | XP_016863357.1 | 3, 4 | 490 | |||
XM_017007869.1 | XP_016863358.1 | 3, 4 | 490 | |||
XM_017007870.1 | XP_016863359.1 | 3, 4 | 490 | |||
XM_017007871.1 | XP_016863360.1 | 3, 4 | 549 | |||
XM_017007873.1 | XP_016863362.1 | 3, 4 | 490 | |||
XM_017007875.1 | XP_016863364.1 | 3, 4 | 449 | |||
GenBank mRNA | AK057729.1 | - | 3, 4 | 396 | ||
AK307317.1 | - | 3, 4 | 449 | |||
BC064600.1 | - | 3, 4 | 454 | 5786393 | ||
BX647575.1 | - | 3, 4 | 501 |